Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 18 - 19 | 2598 | 78 | |
| GenBank mRNA | AF332083.1 | - | 15 - 16 | 2202 | 78 | |
| AF332084.1 | - | 15 - 16 | 2193 | 78 | ||
| AK041296.1 | - | 18 - 19 | 2604 | 78 | ||
| AK076223.1 | - | 18 - 19 | 2579 | 78 | ||
| AK139398.1 | - | 17 - 18 | 3118 | 78 | ||
| AK220467.1 | - | 18 - 19 | 2556 | 78 | ||
| BC064020.1 | - | 6 - 7 | 647 | 78 | ||
| BC068165.1 | - | 18 - 19 | 2530 | 78 | ||
| DQ832283.1 | - | 18 - 19 | 2575 | 78 | ||
| U11812.1 | - | 18 - 19 | 2516 | 78 | ||
| X74438.1 | - | 18 - 19 | 2615 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn
Genomic Map