Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 20 - 21 | 2818 | 83 | |
| GenBank mRNA | AF332083.1 | - | 17 - 18 | 2422 | 83 | |
| AF332084.1 | - | 17 - 18 | 2413 | 83 | ||
| AK041296.1 | - | 20 - 21 | 2824 | 83 | ||
| AK076223.1 | - | 20 - 21 | 2799 | 83 | ||
| AK139398.1 | - | 19 - 20 | 3338 | 83 | ||
| AK220467.1 | - | 20 - 21 | 2776 | 83 | ||
| BC064020.1 | - | 8 - 9 | 867 | 83 | ||
| BC068165.1 | - | 20 - 21 | 2750 | 83 | ||
| DQ832283.1 | - | 20 - 21 | 2795 | 83 | ||
| U11812.1 | - | 20 - 21 | 2736 | 83 | ||
| X74438.1 | - | 20 - 21 | 2835 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn
Genomic Map