Assay Details
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008992.2 | NP_033018.2 | 6 - 7 | 810 | 71 | |
GenBank mRNA | AJ001166.1 | - | 6 - 7 | 714 | 71 | |
AK028420.1 | - | 6 - 7 | 848 | 71 | ||
BC050102.1 | - | 6 - 7 | 918 | 30012556 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
