Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
TC0600003105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_001277330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_001277805.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_008995.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_008995.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_175933.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023943.mm.1 |
NM_175933.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
TC0600003105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_001277330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_001277805.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_008995.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_008995.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_175933.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023942.mm.1 |
NM_175933.2 |
Refseq |