Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_001166406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_001166406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_001166407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_001166407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_009004.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
NM_009004.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002123.mm.1 |
TC1800000291.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_001166406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_001166406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_001166407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_001166407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_009004.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
NM_009004.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002122.mm.1 |
TC1800000291.mm.1 |
MTA TC |