Assay Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177302.1 | NP_001170773.1 | 3 - 4 | 866 | 82 | |
| NM_001177303.1 | NP_001170774.1 | 2 - 3 | 611 | 82 | ||
| NM_009024.2 | NP_033050.2 | 2 - 3 | 733 | 82 | ||
| GenBank mRNA | AB221572.1 | - | 1 - 2 | 180 | 82 | |
| AK042163.1 | - | 2 - 3 | 612 | 82 | ||
| AK154623.1 | - | 2 - 3 | 706 | 82 | ||
| BC010216.1 | - | 2 - 3 | 731 | 3498142 | 82 | |
| BC040383.1 | - | 3 - 4 | 866 | 82 | ||
| BC090396.1 | - | 1 - 2 | 277 | 82 | ||
| CT010231.1 | - | 1 - 2 | 176 | 82 | ||
| S56656.1 | - | 2 - 3 | 713 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rara
Genomic Map