Assay Details
Gene Symbol: Mst1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287261.1 | NP_001274190.1 | 6 - 7 | 921 | 72 | |
NM_009074.2 | NP_033100.2 | 16 - 17 | 3831 | 72 | ||
NR_109782.1 | 16 - 17 | 3862 | 72 | |||
GenBank mRNA | AK053655.1 | - | 16 - 17 | 3845 | 72 | |
BC141379.1 | - | 16 - 17 | 3642 | 9055992 | 72 | |
X74736.1 | - | 16 - 17 | 3831 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
