Assay Details
| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285505.1 | NP_001272434.1 | 8 - 9 | 781 | 61 | |
| NM_001285506.1 | NP_001272435.1 | 7 - 8 | 632 | 61 | ||
| NM_009097.5 | NP_033123.2 | 8 - 9 | 781 | 61 | ||
| GenBank mRNA | AK132856.1 | - | 6 - 7 | 818 | 61 | |
| AK148208.1 | - | 8 - 9 | 774 | 61 | ||
| AK167790.1 | - | 7 - 8 | 619 | 61 | ||
| AK210375.1 | - | 3 - 4 | 264 | 61 | ||
| BC049076.1 | - | 8 - 9 | 732 | 61 | ||
| BC094470.1 | - | 8 - 9 | 734 | 5098650 | 61 | |
| M28489.1 | - | 8 - 9 | 767 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka1
Genomic Map