Assay Details
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159731.1 | NP_001153203.1 | 5 - 6 | 845 | 61 | |
NM_009107.3 | NP_033133.1 | 6 - 7 | 1301 | 61 | ||
GenBank mRNA | BC058401.1 | - | 6 - 7 | 1283 | 5707723 | 61 |
G49237.1 | - | 6 - 7 | 1033 | 61 | ||
S62948.1 | - | 6 - 7 | 1145 | 61 | ||
S62952.1 | - | 5 - 6 | 801 | 61 | ||
X66225.1 | - | 6 - 7 | 1033 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
