| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500007405.mm.1 |
TC0500000841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007405.mm.1 |
NM_009141.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007402.mm.1 |
TC0500000841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007402.mm.1 |
NM_009141.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007403.mm.1 |
TC0500000841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007403.mm.1 |
NM_009141.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007401.mm.1 |
TC0500000841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007401.mm.1 |
NM_009141.3 |
Refseq |