Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009157.4 | NP_033183.1 | 8 - 9 | 933 | 107 | |
GenBank mRNA | AK036928.1 | - | 8 - 9 | 937 | 107 | |
AK044534.1 | - | 8 - 9 | 925 | 107 | ||
AK044698.1 | - | 8 - 9 | 925 | 107 | ||
AK139777.1 | - | 9 - 10 | 956 | 107 | ||
BC029833.1 | - | 6 - 7 | 684 | 3962163 | 107 | |
U18310.1 | - | 8 - 9 | 908 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
