Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500025339.mm.1 |
TC0500002865.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500025339.mm.1 |
NM_001081567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025339.mm.1 |
NM_009158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025334.mm.1 |
TC0500002865.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500025334.mm.1 |
NM_001081567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025334.mm.1 |
NM_009158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025336.mm.1 |
TC0500002865.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500025336.mm.1 |
NM_001081567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025336.mm.1 |
NM_009158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025340.mm.1 |
TC0500002865.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500025340.mm.1 |
NM_001081567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025340.mm.1 |
NM_009158.3 |
Refseq |