Assay Details
| Gene Symbol: Ptk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009184.2 | NP_033210.1 | 4 - 5 | 953 | 93 | |
| GenBank mRNA | AK154717.1 | - | 691 | 93 | ||
| BC125342.1 | - | 4 - 5 | 924 | 40129857 | 93 | |
| BC125344.1 | - | 4 - 5 | 924 | 40129859 | 93 | |
| BC145042.1 | - | 4 - 5 | 893 | 93 | ||
| U16805.1 | - | 4 - 5 | 953 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptk6
Genomic Map