Assay Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009199.2 | NP_033225.1 | 3 - 4 | 396 | 86 | |
| GenBank mRNA | AK018188.1 | - | 3 - 4 | 429 | 86 | |
| AK045632.1 | - | 3 - 4 | 428 | 86 | ||
| AK083097.1 | - | 3 - 4 | 429 | 86 | ||
| BC064797.1 | - | 3 - 4 | 418 | 30029501 | 86 | |
| BC065099.1 | - | 3 - 4 | 420 | 30364069 | 86 | |
| D43797.1 | - | 3 - 4 | 402 | 86 | ||
| U73521.1 | - | 3 - 4 | 396 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a1
Genomic Map