Assay Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009199.2 | NP_033225.1 | 8 - 9 | 951 | 99 | |
| GenBank mRNA | AK018188.1 | - | 8 - 9 | 984 | 99 | |
| AK045632.1 | - | 8 - 9 | 983 | 99 | ||
| AK083097.1 | - | 8 - 9 | 986 | 99 | ||
| BC064797.1 | - | 8 - 9 | 973 | 30029501 | 99 | |
| BC065099.1 | - | 8 - 9 | 975 | 30364069 | 99 | |
| D43797.1 | - | 8 - 9 | 957 | 99 | ||
| U73521.1 | - | 8 - 9 | 951 | 99 | ||
| U75214.1 | - | 4 - 5 | 726 | 99 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF087578.1 | - | Chr. - - [-] on Build | 99 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a1
Genomic Map