Assay Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253892.1 | NP_001240821.1 | 8 - 9 | 1346 | 75 | |
| NM_009207.3 | NP_033233.2 | 9 - 10 | 1542 | 75 | ||
| GenBank mRNA | AK145930.1 | - | 9 - 10 | 1467 | 75 | |
| AK147858.1 | - | 9 - 10 | 1498 | 75 | ||
| BC054102.1 | - | 8 - 9 | 1351 | 30017310 | 75 | |
| J04036.1 | - | 9 - 10 | 1457 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a2
Genomic Map