Assay Details
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113569.1 | NP_001107041.1 | 15 - 16 | 1504 | 74 | |
NM_009295.2 | NP_033321.2 | 15 - 16 | 1499 | 74 | ||
GenBank mRNA | AF326545.1 | - | 15 - 16 | 1358 | 74 | |
AF326563.1 | - | 15 - 16 | 1358 | 74 | ||
BC031728.1 | - | 15 - 16 | 1431 | 4480978 | 74 | |
D45903.1 | - | 15 - 16 | 1393 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
