Assay Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009352.3 | NP_033378.1 | 3 - 4 | 530 | 111 | |
| GenBank mRNA | AB109563.1 | - | 3 - 4 | 510 | 111 | |
| AK021235.1 | - | 3 - 4 | 520 | 111 | ||
| AK036615.1 | - | 3 - 4 | 533 | 111 | ||
| BC034866.1 | - | 3 - 4 | 518 | 3470277 | 111 | |
| U65586.1 | - | 3 - 4 | 510 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Terf1
Genomic Map