Assay Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009367.3 | NP_033393.2 | 1 - 2 | 1564 | 101 | |
| GenBank mRNA | AK159690.1 | - | 1 - 2 | 128 | 101 | |
| BC011170.1 | - | 1 - 2 | 472 | 4035366 | 101 | |
| X57413.1 | - | 1 - 2 | 1563 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfb2
Genomic Map