Assay Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009367.3 | NP_033393.2 | 2 - 3 | 1730 | 63 | |
| GenBank mRNA | AK141552.1 | - | 3 - 4 | 1112 | 63 | |
| AK159690.1 | - | 2 - 3 | 294 | 63 | ||
| BC011170.1 | - | 2 - 3 | 638 | 4035366 | 63 | |
| X57413.1 | - | 2 - 3 | 1729 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfb2
Genomic Map