Assay Details
| Gene Symbol: Tgfbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009370.2 | NP_033396.1 | 2 - 3 | 337 | 71 | |
| GenBank mRNA | AK154381.1 | - | 1 - 2 | 100 | 71 | |
| BC063260.1 | - | 2 - 3 | 337 | 6409765 | 71 | |
| CT010393.1 | - | 2 - 3 | 330 | 71 | ||
| D28526.1 | - | 2 - 3 | 330 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbr1
Genomic Map