Assay Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009373.3 | NP_033399.1 | 5 - 6 | 768 | 68 | |
| GenBank mRNA | AF076928.1 | - | 4 - 5 | 683 | 68 | |
| AF114266.1 | - | 5 - 6 | 733 | 68 | ||
| AK052912.1 | - | 5 - 6 | 743 | 68 | ||
| AK054108.1 | - | 5 - 6 | 743 | 68 | ||
| AK080224.1 | - | 5 - 6 | 768 | 68 | ||
| AK080593.1 | - | 5 - 6 | 743 | 68 | ||
| AK086001.1 | - | 5 - 6 | 743 | 68 | ||
| AK089481.1 | - | 5 - 6 | 771 | 68 | ||
| AK143712.1 | - | 5 - 6 | 804 | 68 | ||
| AK151776.1 | - | 5 - 6 | 769 | 68 | ||
| AK152152.1 | - | 5 - 6 | 743 | 68 | ||
| AK152627.1 | - | 5 - 6 | 769 | 68 | ||
| AK159255.1 | - | 5 - 6 | 743 | 68 | ||
| AK166302.1 | - | 5 - 6 | 743 | 68 | ||
| AK168990.1 | - | 5 - 6 | 744 | 68 | ||
| AK169356.1 | - | 5 - 6 | 771 | 68 | ||
| BC016492.1 | - | 5 - 6 | 709 | 3256943 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm2
Genomic Map