Assay Details
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110265.1 | NP_001103735.1 | 7 - 8 | 885 | 101 | |
NM_001284272.1 | NP_001271201.1 | 7 - 8 | 885 | 101 | ||
NM_009445.2 | NP_033471.2 | 7 - 8 | 885 | 101 | ||
GenBank mRNA | AK041487.1 | - | 7 - 8 | 856 | 101 | |
AK049200.1 | - | 7 - 8 | 101 | |||
AK164093.1 | - | 7 - 8 | 884 | 101 | ||
AX775731.1 | - | 7 - 8 | 851 | 101 | ||
BC058851.1 | - | 7 - 8 | 871 | 30023533 | 101 | |
DV648812.1 | - | 4 - 5 | 371 | 101 | ||
M86377.1 | - | 7 - 8 | 833 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
