Assay Details
| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009506.2 | NP_033532.1 | 4 - 5 | 938 | 137 | |
| GenBank mRNA | AK047844.1 | - | 865 | 137 | ||
| BC096377.1 | - | 4 - 5 | 843 | 5002720 | 137 | |
| U58112.1 | - | 4 - 5 | 834 | 137 | ||
| U73620.1 | - | 4 - 5 | 844 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfc
Genomic Map