Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 7 - 8 | 972 | 69 | |
| GenBank mRNA | AF323659.1 | - | 7 - 8 | 968 | 69 | |
| BC119471.2 | - | 7 - 8 | 920 | 40048166 | 69 | |
| U34920.1 | - | 7 - 8 | 930 | 69 | ||
| Z48745.1 | - | 7 - 8 | 959 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg1
Genomic Map