Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 9 - 10 | 1241 | 77 | |
| GenBank mRNA | AF323659.1 | - | 9 - 10 | 1237 | 77 | |
| BC119471.2 | - | 9 - 10 | 1189 | 40048166 | 77 | |
| CV558133.1 | - | 1 - 2 | 52 | 77 | ||
| U34920.1 | - | 9 - 10 | 1199 | 77 | ||
| Z48745.1 | - | 9 - 10 | 1228 | 77 | ||
More Information
Set Membership: |
|

Abcg1
Genomic Map