Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 12 - 13 | 1616 | 79 | |
| GenBank mRNA | AF323659.1 | - | 12 - 13 | 1612 | 79 | |
| BC119471.2 | - | 12 - 13 | 1564 | 40048166 | 79 | |
| CV558133.1 | - | 4 - 5 | 427 | 79 | ||
| U34920.1 | - | 12 - 13 | 1574 | 79 | ||
| Z48745.1 | - | 12 - 13 | 1603 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg1
Genomic Map