Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
NM_001277255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
NM_001277257.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
NM_001277258.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
NM_001277259.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
NM_009612.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008444.mm.1 |
TC1500001061.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
NM_001277255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
NM_001277257.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
NM_001277258.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
NM_001277259.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
NM_009612.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008445.mm.1 |
TC1500001061.mm.1 |
MTA TC |