Assay Details
| Gene Symbol: Akt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165894.1 | NP_001159366.1 | 5 - 6 | 807 | 68 | |
| NM_009652.3 | NP_033782.1 | 5 - 6 | 807 | 68 | ||
| GenBank mRNA | AK040758.1 | - | 5 - 6 | 613 | 68 | |
| AK154936.1 | - | 5 - 6 | 588 | 68 | ||
| BC066018.1 | - | 5 - 6 | 807 | 5716837 | 68 | |
| M94335.1 | - | 4 - 5 | 459 | 68 | ||
| X65687.1 | - | 5 - 6 | 720 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akt1
Genomic Map