Assay Details
| Gene Symbol: Akt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165894.1 | NP_001159366.1 | 8 - 9 | 1067 | 81 | |
| NM_009652.3 | NP_033782.1 | 8 - 9 | 1067 | 81 | ||
| GenBank mRNA | AK040758.1 | - | 8 - 9 | 873 | 81 | |
| AK154936.1 | - | 8 - 9 | 848 | 81 | ||
| BC066018.1 | - | 8 - 9 | 1067 | 5716837 | 81 | |
| M94335.1 | - | 7 - 8 | 719 | 81 | ||
| X65687.1 | - | 8 - 9 | 980 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akt1
Genomic Map