Assay Details
| Gene Symbol: Cd5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009690.2 | NP_033820.2 | 1 - 2 | 152 | 84 | |
| GenBank mRNA | AF011428.1 | - | 1 - 2 | 58 | 84 | |
| AF018269.1 | - | 1 - 2 | 77 | 84 | ||
| AK159132.1 | - | 1 - 2 | 101 | 84 | ||
| AK159181.1 | - | 1 - 2 | 155 | 84 | ||
| AK159823.1 | - | 1 - 2 | 120 | 84 | ||
| BC006799.1 | - | 1 - 2 | 93 | 3152586 | 84 | |
| BC094459.1 | - | 1 - 2 | 105 | 4161650 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF011428.1,AF018269.1,AK159132.1,AK159181.1,AK159823.1,BC006799.1,BC094459.1,NM_009690.2 transcripts.
Set Membership: |
|

Cd5l
Genomic Map