Assay Details
| Gene Symbol: Atp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009721.6 | NP_033851.1 | 4 - 5 | 1064 | 76 | |
| GenBank mRNA | AK005071.1 | - | 4 - 5 | 711 | 76 | |
| AK010677.1 | - | 4 - 5 | 1065 | 76 | ||
| AK140862.1 | - | 4 - 5 | 711 | 76 | ||
| AK160405.1 | - | 4 - 5 | 710 | 76 | ||
| AK162938.1 | - | 1 - 2 | 53 | 76 | ||
| BC027319.1 | - | 4 - 5 | 703 | 2648940 | 76 | |
| BC094070.1 | - | 4 - 5 | 702 | 4974089 | 76 | |
| X16646.1 | - | 4 - 5 | 601 | 76 | ||
| X61433.1 | - | 3 - 4 | 470 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b1
Genomic Map