Assay Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009739.3 | NP_033869.1 | 2 - 3 | 460 | 57 | |
| GenBank mRNA | AF043070.1 | - | 1 - 2 | 301 | 57 | |
| AK075602.1 | - | 2 - 3 | 386 | 57 | ||
| AK145479.1 | - | 2 - 3 | 431 | 57 | ||
| AK150607.1 | - | 2 - 3 | 452 | 57 | ||
| AK150723.1 | - | 2 - 3 | 452 | 57 | ||
| AK167519.1 | - | 2 - 3 | 452 | 57 | ||
| AK218245.1 | - | 1 - 2 | 169 | 57 | ||
| BC046595.1 | - | 2 - 3 | 430 | 6313106 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bckdk
Genomic Map