Assay Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009739.3 | NP_033869.1 | 9 - 10 | 1111 | 85 | |
| GenBank mRNA | AF043070.1 | - | 8 - 9 | 952 | 85 | |
| AK075602.1 | - | 9 - 10 | 1037 | 85 | ||
| AK145479.1 | - | 9 - 10 | 1082 | 85 | ||
| AK150607.1 | - | 9 - 10 | 1103 | 85 | ||
| AK150723.1 | - | 9 - 10 | 1103 | 85 | ||
| AK167519.1 | - | 9 - 10 | 1103 | 85 | ||
| BC004077.1 | - | 6 - 7 | 539 | 85 | ||
| BC046595.1 | - | 9 - 10 | 1081 | 6313106 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bckdk
Genomic Map