| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015142.mm.1 |
TC0200001927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015144.mm.1 |
TC0200001927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015145.mm.1 |
TC0200001927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015146.mm.1 |
TC0200001927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015141.mm.1 |
TC0200001927.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_009754.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_009754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_207680.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_207680.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_207681.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
NM_207681.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015143.mm.1 |
TC0200001927.mm.1 |
MTA TC |