Assay Details
| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 1 - 2 | 251 | 88 | |
| GenBank mRNA | AF107296.1 | - | 1 - 2 | 206 | 88 | |
| AK133909.1 | - | 1 - 2 | 192 | 88 | ||
| BC031577.1 | - | 1 - 2 | 202 | 4953223 | 88 | |
| BY013487.1 | - | 1 - 2 | 252 | 88 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF107296.1,AK133909.1,BC031577.1,NM_009773.3 transcripts.
Set Membership: |
|

Bub1b
Genomic Map