Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 6 - 7 | 828 | 74 | |
| GenBank mRNA | AK075853.1 | - | 6 - 7 | 798 | 74 | |
| AK088168.1 | - | 6 - 7 | 790 | 74 | ||
| AK128919.1 | - | 6 - 7 | 803 | 74 | ||
| AK133881.1 | - | 6 - 7 | 796 | 74 | ||
| AK148961.1 | - | 6 - 7 | 796 | 74 | ||
| AK150893.1 | - | 6 - 7 | 799 | 74 | ||
| AK159152.1 | - | 6 - 7 | 799 | 74 | ||
| AK159885.1 | - | 6 - 7 | 799 | 74 | ||
| AK159891.1 | - | 6 - 7 | 750 | 74 | ||
| AK159913.1 | - | 6 - 7 | 780 | 74 | ||
| AK169069.1 | - | 6 - 7 | 793 | 74 | ||
| AY040626.1 | - | 6 - 7 | 769 | 74 | ||
| BC013447.1 | - | 6 - 7 | 785 | 4241333 | 74 | |
| BC047126.1 | - | 6 - 7 | 743 | 74 | ||
| L25069.1 | - | 6 - 7 | 800 | 74 | ||
| M62897.1 | - | 6 - 7 | 713 | 74 | ||
| X52108.1 | - | 6 - 7 | 713 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cat
Genomic Map