Assay Details
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009808.4 | NP_033938.3 | 8 - 9 | 1093 | 74 | |
GenBank mRNA | AK030309.1 | - | 8 - 9 | 1068 | 74 | |
AK077256.1 | - | 8 - 9 | 1095 | 74 | ||
AK080045.1 | - | 8 - 9 | 1105 | 74 | ||
AK161525.1 | - | 8 - 9 | 1079 | 74 | ||
AY675223.1 | - | 8 - 9 | 855 | 74 | ||
AY675224.1 | - | 8 - 9 | 1067 | 74 | ||
BC028979.1 | - | 8 - 9 | 1080 | 5375109 | 74 | |
Y13090.1 | - | 7 - 8 | 1083 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
