| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600024772.mm.1 |
TC0600003169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024772.mm.1 |
NM_009829.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024772.mm.1 |
NM_009829.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024774.mm.1 |
TC0600003169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024774.mm.1 |
NM_009829.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024774.mm.1 |
NM_009829.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024776.mm.1 |
TC0600003169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024776.mm.1 |
NM_009829.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024776.mm.1 |
NM_009829.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024771.mm.1 |
TC0600003169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024771.mm.1 |
NM_009829.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024771.mm.1 |
NM_009829.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024775.mm.1 |
TC0600003169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024775.mm.1 |
NM_009829.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024775.mm.1 |
NM_009829.3 |
Refseq |