Assay Details
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009894.3 | NP_034024.2 | 2 - 3 | 316 | 60 | |
GenBank mRNA | AF041377.1 | - | 2 - 3 | 304 | 60 | |
AK003968.1 | - | 2 - 3 | 353 | 60 | ||
BC012664.1 | - | 2 - 3 | 280 | 4218866 | 60 | |
BC046340.1 | - | 2 - 3 | 310 | 6478230 | 60 | |
CB600615.1 | - | 2 - 3 | 293 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619751 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108685 | AF041377.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008056 | AK146687.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008056 | NM_026403.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008056 | AF041377.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008056 | BC012664.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008056 | BC046340.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300768 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292561 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66109 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123896 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008056 | AK146687.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008056 | NM_026403.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008056 | AF041377.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008056 | BC012664.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008056 | BC046340.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P108685 | AF041377.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619717 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619715 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619724 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619726 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123896 | - | - |
Whole Mouse Genome G4122A | A_52_P512365 | AK146687.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P512365 | NM_026403.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P512365 | AF041377.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P512365 | AK003968.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P512365 | BC012664.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108685 | AF041377.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123896 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11217205 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11776225 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11217184 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11217182 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11217210 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11299380 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11776222 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11776228 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11776220 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11776217 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11149818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11776228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11776221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11312303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11312309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11776224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11312304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11312307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619721 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10420166 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420170 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420169 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420167 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420168 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418976_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10420165 | - | - |
Mouse Genome 430 2.0 Array | 1418976_s_at | - | - |
Mouse Genome 430 2.0 Array | 1418976_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12684_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12684_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400016165.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016165.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016165.mm.1 | TC1400002101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016163.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016163.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016163.mm.1 | TC1400002101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016166.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016166.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016166.mm.1 | TC1400002101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016167.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016167.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016167.mm.1 | TC1400002101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016169.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016169.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016169.mm.1 | TC1400002101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016161.mm.1 | NM_009894.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016161.mm.1 | NM_009894.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016161.mm.1 | TC1400002101.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418976_s_at | - | - |
Mouse Expression Set 430 | 1418976_PM_s_at | - | - |
Murine Genome U74v2 Set | 102329_PM_at | - | - |
Murine Genome U74v2 Set | 102329_at | - | - |
Mouse Genome 430A 2.0 Array | 1418976_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1418976_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418976_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418976_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 915343 | - | - |
ABI Mouse Genome Survey Microarray | 925499 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160626 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160626 | - | - |
mouse-6 v1.1 (Illumina) | GI_6753421-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3160626 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160626 | - | - |
