Assay Details
Gene Symbol: Cst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009977.3 | NP_034107.2 | 3 - 4 | 458 | 71 | |
GenBank mRNA | AB015224.1 | - | 3 - 4 | 439 | 71 | |
AF031825.1 | - | 3 - 4 | 387 | 71 | ||
AK004420.1 | - | 3 - 4 | 459 | 71 | ||
AK170583.1 | - | 3 - 4 | 405 | 71 | ||
BC117047.1 | - | 3 - 4 | 443 | 40126366 | 71 | |
BC117049.1 | - | 3 - 4 | 443 | 40126368 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | AK170583.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BC117047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BC117049.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BX630680.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | AK170583.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BC117047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BC117049.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | BX630680.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330847 | AK038716.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330847 | AK129283.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330847 | AK162510.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330847 | XM_001002219.2 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330847 | XM_145521.10 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157018 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510439 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454698 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29709 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | AK170583.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BC117047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BC117049.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BX630680.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | AK170583.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BC117047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BC117049.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | BX630680.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739703 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739690 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739698 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739702 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739682 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739710 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739708 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739699 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739658 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739678 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739715 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739700 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739701 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00526091 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737609 | - | - |
Whole Mouse Genome G4122A | A_52_P915792 | - | - |
Whole Mouse Genome G4122A | A_51_P137419 | AK170583.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BC117047.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BC117049.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BX630680.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | AK170583.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BC117047.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BC117049.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | BX630680.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137419 | NM_009977.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137625 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183856 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183848 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183847 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183854 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183838 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01183843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01773915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01773917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01410041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01410038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01410035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183828 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227551 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01410040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02093761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02034549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02160381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01183839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739712 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01410029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21739716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02184464 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10476946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10476948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10476949 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10476947 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419202_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10476945 | - | - |
Mouse Genome 430 2.0 Array | 1419202_at | - | - |
Mouse Genome 430 2.0 Array | 1419202_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13011_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13011_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200017509.mm.1 | NM_009977.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017509.mm.1 | NM_009977.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017509.mm.1 | TC0200002223.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017510.mm.1 | NM_009977.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017510.mm.1 | NM_009977.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017510.mm.1 | TC0200002223.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017511.mm.1 | NM_009977.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017511.mm.1 | NM_009977.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017511.mm.1 | TC0200002223.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017513.mm.1 | NM_009977.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017513.mm.1 | NM_009977.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017513.mm.1 | TC0200002223.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017512.mm.1 | NM_009977.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017512.mm.1 | NM_009977.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017512.mm.1 | TC0200002223.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419202_PM_at | - | - |
Mouse Expression Set 430 | 1419202_at | - | - |
Murine Genome U74v2 Set | 102638_PM_at | - | - |
Murine Genome U74v2 Set | 102638_at | - | - |
Mouse Genome 430A 2.0 Array | 1419202_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419202_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419202_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419202_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 380369 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 380369 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000380369 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244891 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244891 | - | - |
mouse-6 v1.1 (Illumina) | scl20145.4.1_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 380369 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20145.4.1_7-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380369 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244891 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244891 | - | - |
