Assay Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286396.1 | NP_001273325.1 | 2 - 3 | 241 | 73 | |
| NM_001286397.1 | NP_001273326.1 | 2 - 3 | 251 | 73 | ||
| NM_010018.3 | NP_034148.2 | 2 - 3 | 400 | 73 | ||
| GenBank mRNA | BB843516.1 | - | 2 - 3 | 295 | 73 | |
| BC015269.1 | - | 2 - 3 | 251 | 4239778 | 73 | |
| BC018377.1 | - | 2 - 3 | 241 | 4242375 | 73 | |
| D10210.1 | - | 1 - 2 | 192 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
leucine metabolic process
proline catabolic process
D-serine catabolic process
dopamine biosynthetic process
oxidation-reduction process
D-alanine catabolic process
D-serine metabolic process
D-amino-acid oxidase activity
receptor binding
oxidoreductase activity
oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor
protein dimerization activity
cofactor binding
FAD binding
leucine metabolic process
proline catabolic process
D-serine catabolic process
dopamine biosynthetic process
oxidation-reduction process
D-alanine catabolic process
D-serine metabolic process
D-amino-acid oxidase activity
receptor binding
oxidoreductase activity
oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor
protein dimerization activity
cofactor binding
FAD binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108933 | AK134813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108933 | NM_010018.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100765 | BC018377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100765 | D10210.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100765 | D10211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100765 | M32299.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100765 | AK134813.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41057 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139967 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138765 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100016 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108933 | AK134813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108933 | NM_010018.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100765 | BC018377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100765 | D10210.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100765 | D10211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100765 | M32299.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100765 | AK134813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P452747 | BC018377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P452747 | D10210.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P452747 | D10211.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P452747 | M32299.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P452747 | AK134813.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24001401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139967 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05702535 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100016 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138765 | - | - |
| Whole Mouse Genome G4122A | A_52_P452747 | BC018377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P452747 | D10210.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P452747 | D10211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P452747 | M32299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P452747 | AK134813.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131290 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100765 | BC018377.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100765 | D10210.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100765 | D10211.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100765 | M32299.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100765 | AK134813.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138765 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139967 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100016 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04892199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04892200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04443876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04849492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04849504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04561225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04812485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04736723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04447688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04935259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04115490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04876402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04462415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04673818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04749743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04829365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04876403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10524431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524435 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420709_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10524422 | - | - |
| Mouse Genome 430 2.0 Array | 1420709_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420709_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13142_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13142_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa120_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500010307.mm.1 | TC0500001217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010307.mm.1 | NM_001286396.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010307.mm.1 | NM_001286397.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010307.mm.1 | NM_010018.3 | Refseq |
| Mouse Expression Set 430 | 1420709_s_at | - | - |
| Mouse Expression Set 430 | 1420709_PM_s_at | - | - |
| Murine Genome U74v2 Set | 103602_PM_at | - | - |
| Murine Genome U74v2 Set | 103602_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420709_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420709_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420709_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420709_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 372289 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 60056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 60056 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592844 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592844 | - | - |
| mouse-6 v1.1 (Illumina) | scl0013142.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013142.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2862633 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2862633 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592844 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0013142.1_12-S | - | - |

Dao
Genomic Map