Assay Details
Gene Symbol: Dkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010051.3 | NP_034181.2 | 1 - 2 | 368 | 67 | |
GenBank mRNA | AF030433.1 | - | 1 - 2 | 287 | 67 | |
AK141693.1 | - | 1 - 2 | 392 | 67 | ||
AK144119.1 | - | 1 - 2 | 394 | 67 | ||
BC050189.1 | - | 1 - 2 | 368 | 30022799 | 67 | |
JN966751.1 | - | 1 - 2 | 421 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
