Assay Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010139.3 | NP_034269.2 | 10 - 11 | 1998 | 66 | |
| GenBank mRNA | AK137704.1 | - | 10 - 11 | 1980 | 66 | |
| AK144202.1 | - | 10 - 11 | 1982 | 66 | ||
| BC140960.1 | - | 10 - 11 | 1954 | 9055572 | 66 | |
| U07634.1 | - | 10 - 11 | 1956 | 66 | ||
| X76010.1 | - | 3 - 4 | 213 | 66 | ||
| X78339.1 | - | 10 - 11 | 1960 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha2
Genomic Map