Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800018482.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018482.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018482.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018483.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018483.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018483.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018479.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018479.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018479.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018474.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018474.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018474.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018480.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018480.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018480.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018481.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018481.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018481.mm.1 |
TC0800002422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018478.mm.1 |
NM_010150.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018478.mm.1 |
NM_010150.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018478.mm.1 |
TC0800002422.mm.1 |
MTA TC |