Assay Details
| Gene Symbol: Fcgrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010189.3 | NP_034319.2 | 6 - 7 | 1379 | 72 | |
| GenBank mRNA | AK146933.1 | - | 6 - 7 | 1344 | 72 | |
| L17022.1 | - | 5 - 6 | 1018 | 72 | ||
| Gene Symbol: Mup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC093515.1 | - | 2576 | 72 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642934 | AK011405.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642934 | BC021827.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642934 | XM_915882.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642934 | XM_990993.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95841 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100766 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163002 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161890 | L17022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091186 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01507154 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163002 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100766 | - | - |
| Whole Mouse Genome G4122A | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161890 | L17022.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138122 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163002 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462431 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06605012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462423 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462413 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06462424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06529063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06426724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06415083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06594507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06434377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06462432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06587783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05887809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06529063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06605019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06316384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06605022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06343314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06296810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06450827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06496440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06462429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06343315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05936088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091193 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563087 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416978_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563085 | - | - |
| Mouse Genome 430 2.0 Array | 1416978_at | - | - |
| Mouse Genome 430 2.0 Array | 1416978_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14132_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14132_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700026853.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026853.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026853.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026858.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026858.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026858.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026864.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026864.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026864.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026860.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026860.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026860.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026854.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026854.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026854.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026857.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026857.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026857.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026855.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026855.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026855.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026865.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026865.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026865.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026863.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026863.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026863.mm.1 | TC0700002989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026862.mm.1 | NM_010189.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026862.mm.1 | NM_010189.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026862.mm.1 | TC0700002989.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416978_PM_at | - | - |
| Mouse Expression Set 430 | 1416978_at | - | - |
| Murine Genome U74v2 Set | 97533_at | - | - |
| Murine Genome U74v2 Set | 97533_PM_at | - | - |
| Murine Genome U74v2 Set | 167226_i_at | - | - |
| Murine Genome U74v2 Set | 167226_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416978_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416978_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416978_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416978_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5130315 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130315 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130315 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773706 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773706 | - | - |
| mouse-6 v1.1 (Illumina) | scl31392.8.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130315 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31392.8.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130315 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2917497 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221819 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2917497 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31392.8.1_11-S | - | - |

Fcgrt
Genomic Map