Assay Details
| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010202.5 | NP_034332.2 | 2 - 3 | 553 | 109 | |
| GenBank mRNA | AK082814.1 | - | 2 - 3 | 554 | 109 | |
| BC104312.1 | - | 2 - 3 | 434 | 40044462 | 109 | |
| BC104313.1 | - | 2 - 3 | 434 | 40044466 | 109 | |
| M30642.1 | - | 2 - 3 | 434 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf4
Genomic Map