Assay Details
| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010208.4 | NP_034338.3 | 3 - 4 | 437 | 61 | |
| GenBank mRNA | AK036438.1 | - | 3 - 4 | 359 | 61 | |
| AK036476.1 | - | 3 - 4 | 609 | 61 | ||
| AK038141.1 | - | 3 - 4 | 343 | 61 | ||
| AK089812.1 | - | 3 - 4 | 407 | 61 | ||
| AK155902.1 | - | 3 - 4 | 413 | 61 | ||
| AK157640.1 | - | 391 | 61 | |||
| AK171824.1 | - | 3 - 4 | 391 | 61 | ||
| BC057863.1 | - | 3 - 4 | 306 | 5317449 | 61 | |
| X16440.1 | - | 3 - 4 | 372 | 61 | ||
| X52191.1 | - | 3 - 4 | 344 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgr
Genomic Map