Assay Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010253.3 | NP_034383.1 | 3 - 4 | 287 | 64 | |
| GenBank mRNA | AK133522.1 | - | 3 - 4 | 303 | 64 | |
| BC044055.1 | - | 3 - 4 | 287 | 6476459 | 64 | |
| Z23069.1 | - | 1 - 2 | 66 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gal
Genomic Map