Assay Details
| Gene Symbol: Ghrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010285.2 | NP_034415.1 | 3 - 4 | 282 | 65 | |
| GenBank mRNA | AK005392.1 | - | 3 - 4 | 283 | 65 | |
| AK018796.1 | - | 3 - 4 | 283 | 65 | ||
| AK018816.1 | - | 3 - 4 | 283 | 65 | ||
| AY902312.1 | - | 3 - 4 | 240 | 65 | ||
| BC068204.1 | - | 3 - 4 | 259 | 1496474 | 65 | |
| M31658.1 | - | 3 - 4 | 269 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ghrh
Genomic Map